2-Methoxyethanol, 99+%, extra pure, Thermo Scientific Chemicals
2-Methoxyethanol, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methoxyethanol, 99+%, extra pure, Thermo Scientific Chemicals

2-Methoxyethanol, 99+%, C3H8O2, CAS Number-109-86-4, 2-methoxy-1-ethanol, 3-oxa-1-butanol, dowanol em, egme, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methoxyethanol, methyl cellosolve, methyl oxitol, monomethyl glycol, 2.5L, 99% min. (GC), 01, 467, 124.0 deg.C, 100.0, 99.0, CHEBI:46790
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 180790010
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.90 
Save 8.25 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.90 
Save 8.25 (15%)
Add to cart
2-Methoxyethanol, 99+%, extra pure, Thermo Scientific Chemicals
Catalog number180790010
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.90 
Save 8.25 (15%)
-
Add to cart
Chemical Identifiers
CAS109-86-4
IUPAC Name2-methoxyethan-1-ol
Molecular FormulaC3H8O2
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<15 APHA
Infrared spectrumConforms
Residue after evaporation=<0.005 %
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only