Isopropyl acetate, 99+%, pure, Thermo Scientific Chemicals
Isopropyl acetate, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropyl acetate, 99+%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
2.5 L
25 L
Catalog number 180800010
Price (USD)/ Each
106.65
Online exclusive
118.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
106.65
Online exclusive
118.00 
Save 11.35 (10%)
Add to cart
Isopropyl acetate, 99+%, pure, Thermo Scientific Chemicals
Catalog number180800010
Price (USD)/ Each
106.65
Online exclusive
118.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS108-21-4
IUPAC Namepropan-2-yl acetate
Molecular FormulaC5H10O2
InChI KeyJMMWKPVZQRWMSS-UHFFFAOYSA-N
SMILESCC(C)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Water=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only