Propionitrile, 99%, extra pure
Propionitrile, 99%, extra pure
Propionitrile, 99%, extra pure
Propionitrile, 99%, extra pure
Thermo Scientific Chemicals

Propionitrile, 99%, extra pure

CAS: 107-12-0 | C3H5N | 55.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 180891000
Price (USD)
60.65
Online Exclusive
67.40
Save 6.75 (10%)
Each
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
60.65
Online Exclusive
67.40
Save 6.75 (10%)
Each
Add to cart
Chemical Identifiers
CAS107-12-0
IUPAC Namepropanenitrile
Molecular FormulaC3H5N
InChI KeyFVSKHRXBFJPNKK-UHFFFAOYSA-N
SMILESCCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.3650 to 1.3680 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link