Naphthalene, 99%
Naphthalene, 99%
Naphthalene, 99%
Thermo Scientific Chemicals

Naphthalene, 99%

CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 180900025
Price (EUR)
95,20
Online Exclusive
112,00
Save 16,80 (15%)
Each
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
95,20
Online Exclusive
112,00
Save 16,80 (15%)
Each
Add to cart
Chemical Identifiers
CAS91-20-3
IUPAC Namenaphthalene
Molecular FormulaC10H8
InChI KeyUFWIBTONFRDIAS-UHFFFAOYSA-N
SMILESC1=CC2=CC=CC=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder or crystals or flakes or
Appearance (Form)tablets or balls
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473923Certificate of AnalysisJun 27, 2025180902500, 180900025, 180900010, 180900050
A0468034Certificate of AnalysisMay 17, 2025180902500, 180900025, 180900010, 180900050
A0465623Certificate of AnalysisAug 22, 2024180902500, 180900025, 180900010, 180900050
A0461639Certificate of AnalysisApr 05, 2024180902500, 180900025, 180900010, 180900050
A0461638Certificate of AnalysisApr 05, 2024180902500, 180900025, 180900010, 180900050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable solids
Category 2
Acute oral toxicity
Category 4
Carcinogenicity
Category 2
Acute aquatic toxicity
Category 1
Chronic aquatic toxicity
Category 1
Label Elements
Signal Word

Warning

Hazard Statements

H302 - Harmful if swallowed

H351 - Suspected of causing cancer

H410 - Very toxic to aquatic life with long lasting effects

Physical Hazards

H228 - Flammable solid

Precautionary Statements

P201 - Obtain special instructions before use

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P264 - Wash face, hands and any exposed skin thoroughly after handling

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P312 - Call a POISON CENTER or doctor if you feel unwell

P370 + P378 - In case of fire: Use dry sand, dry chemical or alcohol-resistant foam for extinction

Share catalog number, name or link

1x1 image pixel for data collection