The BioPrime™ DNA Labeling System is specifically designed for use in the preparation of biotinylated probes. Random primers (octamers) are annealed to the denatured DNA template and extended by Klenow fragment in the presence of biotin-14-dCTP to produce sensitive biotinylated-DNA probes for use in the nonradioactive detection of DNA and RNA. With the BioPrime™ DNA Labeling System, considerable net DNA synthesis occurs resulting in a 10–40 fold amplification of the probe. This product is particularly well suited for situations in which the DNA is limited. Probes made with this product have been used for hybridization to Southern and northern blots, plaque and colony lifts, and for in situ hybridization to chromosome spreads. The BioPrime™ DNA Labeling System:
• Requires less template DNA than nick translation • Amplifies template to provide an increased amount of biotinylated probe • Labels 50 to 500 ng of template DNA in one reaction
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection Method
Biotin-based
Label or Dye
Biotin
Final Product Type
Probes (Labeled DNA)
Labeling Target
DNA (General), Genomic DNA
Includes Label or Dye
Yes
Labeling Method
Direct Labeling
Product Line
BioPrime™
Product Type
DNA Labeling System
Quantity
30 Reactions
Shipping Condition
Approved for shipment on Wet or Dry Ice
Contents & Storage
• 700 μl 2.5X Random Primers Solution • 175 μl 10X dNTP Mixture • 100 μg Control DNA • 35 μl Klenow Fragment (40 U/μl) • 500 μl Stop Buffer • 1.25 ml Distilled Water