Thermo Scientific Chemicals

2-Chlorophenol, 98+%, Thermo Scientific Chemicals

Catalog number: 180991000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Chlorophenol, 98+%, Thermo Scientific Chemicals

Catalog number: 180991000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 180991000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
95-57-8
IUPAC Name
2-chlorophenol
Molecular Formula
C6H5ClO
InChI Key
ISPYQTSUDJAMAB-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1Cl
Appearance (Color)
Clear light yellow to brown
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Refractive index
1.5580 to 1.5600 (20°C, 589 nm)
GC
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text