Thermo Scientific Chemicals

Cyanuric acid, 98%, Thermo Scientific Chemicals

Catalog number: 181020010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Cyanuric acid, 98%, Thermo Scientific Chemicals

Catalog number: 181020010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 181020010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
108-80-5
IUPAC Name
1,3,5-triazinane-2,4,6-trione
Molecular Formula
C3H3N3O3
InChI Key
ZFSLODLOARCGLH-UHFFFAOYSA-N
SMILES
O=C1NC(=O)NC(=O)N1
Appearance (Color)
White to light beige
Appearance (Form)
Crystalline powder and/or lumps
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text