Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dicyclohexylamine, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 181051000
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
40.80 
Save 6.15 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
40.80 
Save 6.15 (15%)
Add to cart
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Catalog number181051000
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
40.80 
Save 6.15 (15%)
-
Add to cart
Chemical Identifiers
IUPAC NameN-cyclohexylcyclohexanamine
Molecular FormulaC12H23N
InChI KeyXBPCUCUWBYBCDP-UHFFFAOYSA-N
SMILESC1CCC(CC1)NC1CCCCC1
Molecular Weight (g/mol)181.32
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Refractive index1.4830 to 1.4850 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only