Thermo Scientific Chemicals

n-Butyllithium, 1.6M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 181275000
500 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

n-Butyllithium, 1.6M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 181275000
500 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 181275000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-72-8
IUPAC Name
butyllithium
Molecular Formula
C4H9Li
InChI Key
MZRVEZGGRBJDDB-UHFFFAOYSA-N
SMILES
[Li]CCCC
Free base
=<1 %
Appearance (Color)
Clear to hazy colorless to yellow
Appearance
May develop some turbidity or precipitate
Appearance (Form)
Solution
Titration Active base
14.0 to 16.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text