Thermo Scientific Chemicals

Dimethyl adipate, 99+%, Thermo Scientific Chemicals

Catalog number: 181315000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Dimethyl adipate, 99+%, Thermo Scientific Chemicals

Catalog number: 181315000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 181315000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
627-93-0
IUPAC Name
1,6-dimethyl hexanedioate
Molecular Formula
C8H14O4
InChI Key
UDSFAEKRVUSQDD-UHFFFAOYSA-N
SMILES
COC(=O)CCCCC(=O)OC
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=99.0 %
Refractive index
1.4270 to 1.4300 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text