5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals
5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 181331000
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
Add to cart
5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals
Catalog number181331000
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS99-31-0
IUPAC Name5-aminobenzene-1,3-dicarboxylic acid
Molecular FormulaC8H7NO4
InChI KeyKBZFDRWPMZESDI-UHFFFAOYSA-N
SMILESNC1=CC(=CC(=C1)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to light brown
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only