Thermo Scientific Chemicals

Methyl acetate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 181380025
2.5 L, Glass Bottle, Each
Thermo Scientific Chemicals

Methyl acetate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 181380025
2.5 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 181380025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-20-9
IUPAC Name
methyl acetate
Molecular Formula
C3H6O2
InChI Key
KXKVLQRXCPHEJC-UHFFFAOYSA-N
SMILES
COC(C)=O
Appearance (Form)
Clear liquid
Color scale
=<10 APHA
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.3600 to 1.3630 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text