1x1 image pixel for data collection
L-Nicotine, 99+%
L-Nicotine, 99+%
L-Nicotine, 99+%
Thermo Scientific Chemicals

L-Nicotine, 99+%

L-Nicotine, CAS # 54-11-5, has been shown in laboratory studies to alleviate smoking habits. It is also known as 3-(1-methyl-2-pyrrolidinyl)pyridine or (S)-(-)-nicotine. | CAS: 54-11-5 | C10H14N2 | 162.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
181425000500 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 181425000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS54-11-5
IUPAC Name3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
Molecular FormulaC10H14N2
InChI KeySNICXCGAKADSCV-JTQLQIEISA-N
SMILESCN1CCC[C@H]1C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
GC>=99.0 %
Specific optical rotationon anhydrous basis
Specific optical rotation-140° to -152° (20°C, 589 nm) (c=2,absolute ethanol)
Refractive index1.5255 to 1.5285 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L-Nicotine is a naturally occuring alkaloid in the nightshade family of plants
  • It is also known as 3-(1-methyl-2-pyrrolidinyl)pyridine or (S)-(-)-nicotine

Applications

  • L-Nicotine has been shown in studies to alleviate smoking habits
  • It has also been shown to enhance cognitive functions such as memory
  • Studies show that lower levels of nicotine in the form of gum may also help eliminate smoking habits
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link