Thermo Scientific Chemicals

N,N-Dimethyl-p-phenylenediamine sulfate, 99%, Thermo Scientific Chemicals

Catalog number: 181430250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N-Dimethyl-p-phenylenediamine sulfate, 99%, Thermo Scientific Chemicals

Catalog number: 181430250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 181430250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
536-47-0
IUPAC Name
N1,N1-dimethylbenzene-1,4-diamine; sulfuric acid
Molecular Formula
C8H14N2O4S
InChI Key
GLUKPDKNLKRLHX-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.CN(C)C1=CC=C(N)C=C1
Appearance (Form)
Powder or crystals and/or chunks
Titration with NaOH
>=98.5 %
Appearance (Color)
White to grey to brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text