Thermo Scientific Chemicals

18-Crown-6, 99%, Thermo Scientific Chemicals

Catalog number: 181565000
500 g, Glass Bottle, Each
Thermo Scientific Chemicals

18-Crown-6, 99%, Thermo Scientific Chemicals

Catalog number: 181565000
500 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 181565000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
17455-13-9
Appearance (Color)
Clear colorless to white
Infrared spectrum
Conforms
Appearance (Form)
Crystals or crystalline mass or liquid
Melting point
38°C to 42°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text