Informational:Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Informational:BioProbes™ Journal of Cell Biology Applications—Subscribe today »
Invitrogen™

Nick Translation System

Catalog number: 18160010
Have Questions?
Invitrogen™

Nick Translation System

Catalog number: 18160010
Have Questions?
Catalog Number
18160010
Unit Size
50 rxns
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1816001050 rxns
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Nick Translation System is ideal for both radioactive and non-radioactive labeling of DNA. The Nick Translation System:

• Provides five pre-mixed nucleotide solutions for flexibility in labeled nucleotide incorporation
• Yields >108 cpm/μg control DNA using [ - 32 P]-dCTP
• Labels 1 μg of DNA in one reaction
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Radioactive, Fluorescence
Final Product Type
Probes (Labeled DNA)
Labeling Target
DNA (General)
Includes Label or Dye
No
Labeling Method
Direct Labeling
No. of Reactions
50
Product Type
Nick Translation System
Quantity
50 Reactions
Shipping Condition
Wet Ice

Contents & Storage

The Nick Translation System includes an enzyme mix containing DNA Polymerase I and DNase I, dNTPs, control DNA, stop buffer, and distilled water. Store at -20°C. Guaranteed stable for 6 months when properly stored.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text