Orion™ 1816DO Low-Level Dissolved Oxygen Analyzer
Orion™ 1816DO Low-Level Dissolved Oxygen Analyzer
Actual product may vary
Thermo Scientific™
Orion™ 1816DO Low-Level Dissolved Oxygen Analyzer
Continuously monitor oxygen across a wide variety of low and trace level ppb level applications with the Thermo Scientific™ Orion™ 1816DO Low-Level Dissolved Oxygen Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
1816FPDO Fluidics Panel
1816D2DO Monitor without Sensor
1816D0DO Monitor
Catalog number 1816FP
Price (USD)
-
Request A Quote
Type:
DO Fluidics Panel
Continuously monitor oxygen across a wide variety of low and trace level ppb level applications with the Thermo Scientific™ Orion™ 1816DO Low-Level Dissolved Oxygen Analyzer. Offering highly accurate measurement performance with proven reliability, the 1816DO analyzer provides effective process control you can trust.
  • PTFE membrane caps for sensor refurbishment in minutes
  • Rugged and sensitive galvanic dissolved oxygen sensor with rapid response
  • Offers high performance results
  • Galvanic dissolved oxygen sensor with unique Guard Ring™ technology; capable of consuming oxygen and associated gases
Specifications
Description1816 Trace DO Fluidics Panel Only
TypeDO Fluidics Panel
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection