Thermo Scientific Chemicals

4-Bromobenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 181770500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromobenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 181770500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 181770500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
873-75-6
IUPAC Name
(4-bromophenyl)methanol
Molecular Formula
C7H7BrO
InChI Key
VEDDBHYQWFOITD-UHFFFAOYSA-N
SMILES
OCC1=CC=C(Br)C=C1
Melting point
75°C to 78°C
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text