Bis(trimethylsilyl)acetylene, 99%, Thermo Scientific Chemicals
Bis(trimethylsilyl)acetylene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(trimethylsilyl)acetylene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 182010100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nametrimethyl[2-(trimethylsilyl)ethynyl]silane
Molecular FormulaC8H18Si2
InChI KeyZDWYFWIBTZJGOR-UHFFFAOYSA-N
SMILESC[Si](C)(C)C#C[Si](C)(C)C
Molecular Weight (g/mol)170.40
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid after melting
GC>=98.5 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4260 to 1.4280 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only