2-Methoxyphenethylamine, 98%, Thermo Scientific Chemicals
2-Methoxyphenethylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methoxyphenethylamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 182171000
Price (USD)/ Each
322.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
322.00
Add to cart
2-Methoxyphenethylamine, 98%, Thermo Scientific Chemicals
Catalog number182171000
Price (USD)/ Each
322.00
-
Add to cart
Chemical Identifiers
IUPAC Name2-(2-methoxyphenyl)ethan-1-aminium
Molecular FormulaC9H14NO
InChI KeyWSWPCNMLEVZGSM-UHFFFAOYSA-O
SMILESCOC1=CC=CC=C1CC[NH3+]
Molecular Weight (g/mol)152.22
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to light brown
Refractive index1.5400 to 1.5420 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with HClO4>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only