2,5-Dimethoxybenzyl alcohol, 99%, Thermo Scientific Chemicals
2,5-Dimethoxybenzyl alcohol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,5-Dimethoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 182200050
Price (USD)/ Each
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(2,5-dimethoxyphenyl)methanol
Molecular FormulaC9H12O3
InChI KeyWGQMUABRZGUAOS-UHFFFAOYSA-N
SMILESCOC1=CC=C(OC)C(CO)=C1
Molecular Weight (g/mol)168.19
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.5460 to 1.5480 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only