Thermo Scientific Chemicals

2,5-Dimethoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 182200250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

2,5-Dimethoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 182200250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 182200250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
33524-31-1
IUPAC Name
(2,5-dimethoxyphenyl)methanol
Molecular Formula
C9H12O3
InChI Key
WGQMUABRZGUAOS-UHFFFAOYSA-N
SMILES
COC1=CC=C(OC)C(CO)=C1
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Viscous liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.5460 to 1.5480 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text