Thermo Scientific Chemicals

Cyclobutanemethanol, 99%, Thermo Scientific Chemicals

Catalog number: 182230250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Cyclobutanemethanol, 99%, Thermo Scientific Chemicals

Catalog number: 182230250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 182230250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4415-82-1
IUPAC Name
cyclobutylmethanol
Molecular Formula
C5H10O
InChI Key
WPOPOPFNZYPKAV-UHFFFAOYSA-N
SMILES
OCC1CCC1
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.4450 to 1.4470 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text