Methyl cellulose, viscosity 15 cP (2% solution in water)
Methyl cellulose, viscosity 15 cP (2% solution in water)
Methyl cellulose, viscosity 15 cP (2% solution in water)
Methyl cellulose, viscosity 15 cP (2% solution in water)
Thermo Scientific Chemicals

Methyl cellulose, viscosity 15 cP (2% solution in water)

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number 182312500
Price (USD)
80.65
Online Exclusive
89.10
Save 8.45 (9%)
Each
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
80.65
Online Exclusive
89.10
Save 8.45 (9%)
Each
Add to cart
Chemical Identifiers
CAS9004-67-5
IUPAC Name3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
Molecular FormulaC20H38O11
InChI KeyYLGXILFCIXHCMC-UHFFFAOYNA-N
SMILESCOCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Loss on drying=<5 %
Viscosity12 to 18 cP (2 % in water at 20°C)
Appearance (Color)White to yellowish-white to greyish-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link