Chemical Identifiers
CAS2411-89-4
IUPAC Name2-[({5-[1-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid
Molecular FormulaC32H32N2O12
InChI KeyIYZPEGVSBUNMBE-UHFFFAOYSA-N
SMILESCC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1
View more