Thermo Scientific Chemicals

o-Cresolphthalein Complexone, pure

Catalog number: 182441000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

o-Cresolphthalein Complexone, pure

Catalog number: 182441000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 182441000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2411-89-4
IUPAC Name
2-[({5-[1-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid
Molecular Formula
C32H32N2O12
InChI Key
IYZPEGVSBUNMBE-UHFFFAOYSA-N
SMILES
CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1
Appearance (Color)
Off-white to pink-beige
UV spectrum
Complies
Loss on drying
=<10 % (1 g, 105°C)
Sulfated ash
=<0.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text