Tosylmethyl isocyanide, 98%
Tosylmethyl isocyanide, 98%
Tosylmethyl isocyanide, 98%
Tosylmethyl isocyanide, 98%
Thermo Scientific Chemicals

Tosylmethyl isocyanide, 98%

CAS: 36635-61-7 | C9H9NO2S | 195.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number 182451000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS36635-61-7
IUPAC Name1-isocyanomethanesulfonyl-4-methylbenzene
Molecular FormulaC9H9NO2S
InChI KeyCFOAUYCPAUGDFF-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)C[N+]#[C-]
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point109°C to 115°C (Dec)
Appearance (Form)Powder and/or chunks
Appearance (Color)Light yellow to beige to light brown
Infrared spectrumConforms
HPLC>=97.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection