Informational:Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Informational:BioProbes™ Journal of Cell Biology Applications—Subscribe today »
Invitrogen™

BioNick™ DNA Labeling System

Catalog number: 18247015
Have Questions?
Invitrogen™

BioNick™ DNA Labeling System

Catalog number: 18247015
Have Questions?
Catalog Number
18247015
Unit Size
50 rxns
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1824701550 rxns
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The BioNick™ DNA Labeling System is ideal for generating biotinylated DNA probes by nick translation optimized for use in non-radioactive in situ hybridizations. These probes can also be used in Southern or northern blots, plaque lifts, colony hybridizations, and dot blot hybridizations. Using the BioNick™ DNA Labeling System:

• DNA is labeled with biotin-14-dATP, producing probe sizes from 50 to 500 bp
• One reaction labels 1 μg of template DNA
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Biotin-based
Label or Dye
Biotin
Final Product Type
Probes (Labeled DNA)
Labeling Target
DNA (General)
Includes Label or Dye
Yes
Labeling Method
Direct Labeling
No. of Reactions
50
Product Line
BioNick™
Product Type
DNA Labeling System
Quantity
50 Reactions
Shipping Condition
Approved for shipment on Wet or Dry Ice

Contents & Storage

The BioNick™ DNA Labeling System includes 10X dNTP mix containing biotin-14-dATP, 10X enzyme mix containing DNA Polymerase I and DNase I, control DNA, stop buffer, and distilled water. Store at -20°C. Guaranteed stable for 6 months when properly stored.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text