Thermo Scientific Chemicals

2-Mercaptopyridine-N-oxide, 99%, Thermo Scientific Chemicals

Catalog number: 182670050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Mercaptopyridine-N-oxide, 99%, Thermo Scientific Chemicals

Catalog number: 182670050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 182670050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1121-31-9
IUPAC Name
1-hydroxy-1,2-dihydropyridine-2-thione
Molecular Formula
C5H5NOS
InChI Key
YBBJKCMMCRQZMA-UHFFFAOYSA-N
SMILES
ON1C=CC=CC1=S
Appearance (Color)
Off-white to yellow to yellow-green to beige to
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals or chunks
Titration Iodometric
>=98.5 %
Appearance (Color)
grey-green

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text