182702 Full specifications
Case of 20 850 cm2 Smooth 2/Pk., 20/Case - Bottle Surface Smooth Color Clear For Use With (Application) Cell Culture Expansion, Vaccine Production, Antibody Production Material Polystyrene No. per Case 20 No. per Pack 2 Packaging 2/Pk., 20/Case Sterility Sterile Cell Growth Area 850 cm2 Closure Included Yes Graduated Yes Shape Round Product Type Roller Bottle Closure Type Easy on/off, Non-vented Cap Unit Size Case of 20
181702 Full specifications
Case of 20 850 cm2 Smooth 2/Pk., 20/Case - Bottle Surface Smooth Color Clear For Use With (Application) Cell Culture Expansion, Vaccine Production, Antibody Production Material Polystyrene No. per Case 20 No. per Pack 2 Packaging 2/Pk., 20/Case Sterility Sterile Cell Growth Area 850 cm2 Closure Included Yes Graduated Yes Shape Round Product Type Roller Bottle Closure Type Easy on/off, Vented Cap Unit Size Case of 20
182720 Full specifications
Case of 20 850 cm2 Smooth 20/Pk., 20/Case - Bottle Surface Smooth Color Clear For Use With (Application) Cell Culture Expansion, Vaccine Production, Antibody Production Material Polystyrene No. per Case 20 No. per Pack 20 Packaging 20/Pk., 20/Case Sterility Sterile Cell Growth Area 850 cm2 Closure Included Yes Graduated Yes Shape Round Product Type Roller Bottle Closure Type Easy on/off, Non-vented Cap Unit Size Case of 20
182744 Full specifications
Case of 20 850 cm2 Smooth 20/Pk., 20/Case; Double-bagged - Bottle Surface Smooth Color Clear For Use With (Application) Cell Culture Expansion, Vaccine Production, Antibody Production Material Polystyrene No. per Case 20 No. per Pack 20 Packaging 20/Pk., 20/Case; Double-bagged Sterility Sterile Cell Growth Area 850 cm2 Closure Included Yes Graduated Yes Shape Round Product Type Roller Bottle Closure Type Easy on/off, Non-vented Cap Unit Size Case of 20
183302 Full specifications
Case of 20 850 cm2 Smooth surface, shallow indent 2/Pk., 20/Case - Bottle Surface Smooth surface, shallow indent Color Clear For Use With (Application) Cell Culture Expansion, Vaccine Production, Antibody Production Material Polystyrene No. per Case 20 No. per Pack 2 Packaging 2/Pk., 20/Case Sterility Sterile Cell Growth Area 850 cm2 Closure Included Yes Graduated Yes Shape Round Product Type Roller Bottle Closure Type EZ Twist Vented Cap Unit Size Case of 20
183902 Full specifications
Case of 20 850 cm2 Smooth surface, deep indent 2/Pk., 20/Case - Bottle Surface Smooth surface, deep indent Color Clear For Use With (Application) Cell Culture Expansion, Vaccine Production, Antibody Production Material Polystyrene No. per Case 20 No. per Pack 2 Packaging 2/Pk., 20/Case Sterility Sterile Cell Growth Area 850 cm2 Graduations Yes Closure Included Yes Graduated Yes Mouth Narrow Shape Round Product Type Roller Bottle Closure Material HDPE Closure Type EZ Twist Vented Cap Unit Size Case of 20
184302 Full specifications
Case of 20 850 cm2 Smooth surface, shallow indent 2/Pk., 20/Case - Bottle Surface Smooth surface, shallow indent Color Clear For Use With (Application) Cell Culture Expansion, Vaccine Production, Antibody Production Material Polystyrene No. per Case 20 No. per Pack 2 Packaging 2/Pk., 20/Case Sterility Sterile Cell Growth Area 850 cm2 Graduations Yes Closure Included Yes Graduated Yes Mouth Narrow Shape Round Product Type Roller Bottle Closure Material HDPE Closure Type EZ Twist Non-vented Cap Unit Size Case of 20
184344 Full specifications
Case of 20 850 cm2 Smooth surface, shallow indent 20/Pk., 20/Case; Double-bagged - Bottle Surface Smooth surface, shallow indent Color Clear For Use With (Application) Cell Culture Expansion, Vaccine Production, Antibody Production Material Polystyrene No. per Case 20 No. per Pack 20 Packaging 20/Pk., 20/Case; Double-bagged Sterility Sterile Cell Growth Area 850 cm2 Graduations Yes Closure Included Yes Graduated Yes Mouth Narrow Shape Round Product Type Roller Bottle Closure Material HDPE Closure Type EZ Twist Non-vented Cap Unit Size Case of 20
184902 Full specifications
Case of 20 850 cm2 Smooth surface, deep indent 2/Pk., 20/Case; Double-bagged - Bottle Surface Smooth surface, deep indent Color Clear For Use With (Application) Cell Culture Expansion, Vaccine Production, Antibody Production Material Polystyrene No. per Case 20 No. per Pack 2 Packaging 2/Pk., 20/Case; Double-bagged Sterility Sterile Cell Growth Area 850 cm2 Graduations Yes Closure Included Yes Graduated Yes Mouth Narrow Shape Round Product Type Roller Bottle Closure Material HDPE Closure Type EZ Twist Non-vented Cap Unit Size Case of 20
184920 Full specifications
Case of 20 850 cm2 Smooth surface, deep indent 20/Pk., 20/Case - Bottle Surface Smooth surface, deep indent Color Clear For Use With (Application) Cell Culture Expansion, Vaccine Production, Antibody Production Material Polystyrene No. per Case 20 No. per Pack 20 Packaging 20/Pk., 20/Case Sterility Sterile Cell Growth Area 850 cm2 Graduations Yes Closure Included Yes Graduated Yes Mouth Narrow Shape Round Product Type Roller Bottle Closure Material HDPE Closure Type EZ Twist Non-vented Cap Unit Size Case of 20