Trimethylene sulfide, Thermo Scientific Chemicals
Product Image
Thermo Scientific Chemicals

Trimethylene sulfide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 182910050
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.50 
Save 13.85 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.50 
Save 13.85 (15%)
Add to cart
Trimethylene sulfide, Thermo Scientific Chemicals
Catalog number182910050
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.50 
Save 13.85 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namethietane
Molecular FormulaC3H6S
InChI KeyXSROQCDVUIHRSI-UHFFFAOYSA-N
SMILESC1CSC1
Molecular Weight (g/mol)74.14
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.5040 to 1.5140 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only