1-Phenyl-2-propanol, 98%, Thermo Scientific Chemicals
1-Phenyl-2-propanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Phenyl-2-propanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 183060250
Price (USD)/ Each
140.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
165.00 
Save 24.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
140.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
165.00 
Save 24.35 (15%)
Add to cart
1-Phenyl-2-propanol, 98%, Thermo Scientific Chemicals
Catalog number183060250
Price (USD)/ Each
140.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
165.00 
Save 24.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1-phenylpropan-2-ol
Molecular FormulaC9H12O
InChI KeyWYTRYIUQUDTGSX-UHFFFAOYNA-N
SMILESCC(O)CC1=CC=CC=C1
Molecular Weight (g/mol)136.19
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to almost colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5200 to 1.5240 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only