Thermo Scientific Chemicals

Poly(ethylene oxide), M.W. 600,000 to 1,000,000, Thermo Scientific Chemicals

Catalog number: 183225000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Poly(ethylene oxide), M.W. 600,000 to 1,000,000, Thermo Scientific Chemicals

Catalog number: 183225000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 183225000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
25322-68-3
Molecular Formula
(C2H4O)n
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES
[H]OCCO
Appearance (Form)
Powder
Loss on drying
=<2 % (10 g, 5 h, 10 mmHg)
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Viscosity
300 to 600 mPa.s (2 % w/w in water at 25°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text