Poly(vinyl chloride), carboxylated, aver. M.W.220000, aver. carboxyl content 1.8 wt.%, Thermo Scientific Chemicals
Poly(vinyl chloride), carboxylated, aver. M.W.220000, aver. carboxyl content 1.8 wt.%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(vinyl chloride), carboxylated, aver. M.W.220000, aver. carboxyl content 1.8 wt.%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Packaging:
Glass bottle
Catalog number 183312500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
Molecular Formula(C2H3R)n
SMILES*-CC(-*)[*]
MDL NumberMFCD00084460
CAS25702-80-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only