Thermo Scientific Chemicals

Poly(vinyl alcohol), 95.5-96.5% hydrolyzed, M.W. approx. 85,000-124,000, Thermo Scientific Chemicals

Catalog number: 183391000
100 g, Each
Thermo Scientific Chemicals

Poly(vinyl alcohol), 95.5-96.5% hydrolyzed, M.W. approx. 85,000-124,000, Thermo Scientific Chemicals

Catalog number: 183391000
100 g, Each
Quantity
Catalog number: 183391000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9002-89-5
Molecular Formula
(C2H4O)n
SMILES
OC(-*)C-*
MDL Number
MFCD00081922
Appearance (Color)
White to cream
Infrared spectrum
Conforms
Viscosity
27 to 31 cP (4 % in water)
Appearance (Form)
Granular powder
Volatile matter
=<5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text