Thermo Scientific Chemicals

3-Nitrobenzamide, 98%, Thermo Scientific Chemicals

Catalog number: 183441000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Nitrobenzamide, 98%, Thermo Scientific Chemicals

Catalog number: 183441000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 183441000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
645-09-0
IUPAC Name
3-nitrobenzamide
Molecular Formula
C7H6N2O3
InChI Key
KWAYEPXDGHYGRW-UHFFFAOYSA-N
SMILES
NC(=O)C1=CC=CC(=C1)[N+]([O-])=O
Appearance (Color)
Yellow
Infrared spectrum
Conforms
HPLC
>=97.5 %
Appearance (Form)
Crystalline powder or crystals
Melting point
140°C to 144°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text