Thermo Scientific Chemicals

Pyridinium chlorochromate, 98%, Thermo Scientific Chemicals

Catalog number: 183675000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Pyridinium chlorochromate, 98%, Thermo Scientific Chemicals

Catalog number: 183675000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 183675000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26299-14-9
IUPAC Name
hydrogen (trioxochromio)-λ³-chloranidyl pyridine
Molecular Formula
C5H6ClCrNO3
InChI Key
BUCMHZVAGRYTNK-UHFFFAOYSA-O
SMILES
[H+].[Cl-][Cr](=O)(=O)=O.C1=CC=NC=C1
Appearance (Color)
Yellow to orange
Appearance (Form)
Powder or crystals and/or chunks
Infrared spectrum
Conforms
Titration Iodometric
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text