Thermo Scientific Chemicals

Isopropanol, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 184130100
10 L, Plastic Drum, Each
Thermo Scientific Chemicals

Isopropanol, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 184130100
10 L, Plastic Drum, Each
Quantity
Packaging
Catalog number: 184130100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
67-63-0
IUPAC Name
propan-2-ol
Molecular Formula
C3H8O
InChI Key
KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES
CC(C)O
Color scale
=<10 APHA
Water
=<0.1 % (Coulometric)
Residue after evaporation
=<0.001 %
Copper (Cu)
=<0.05 ppm
Magnesium (Mg)
=<0.1 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text