Trichloroacetic acid, sodium salt, 97%, Thermo Scientific Chemicals
Trichloroacetic acid, sodium salt, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trichloroacetic acid, sodium salt, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 184155000
Price (USD)/ Each
106.00
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
106.00
Add to cart
Trichloroacetic acid, sodium salt, 97%, Thermo Scientific Chemicals
Catalog number184155000
Price (USD)/ Each
106.00
-
Add to cart
Chemical Identifiers
CAS650-51-1
IUPAC Namesodium trichloroacetate
Molecular FormulaC2Cl3NaO2
InChI KeySAQSTQBVENFSKT-UHFFFAOYSA-M
SMILES[Na+].[O-]C(=O)C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Powder or granular powder
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only