7-Aminocephalosporanic acid, 95-102%, Thermo Scientific Chemicals
7-Aminocephalosporanic acid, 95-102%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Aminocephalosporanic acid, 95-102%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 184410010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS957-68-6
IUPAC Name(6R,7R)-3-[(acetyloxy)methyl]-7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Molecular FormulaC10H12N2O5S
InChI KeyHSHGZXNAXBPPDL-HZGVNTEJSA-N
SMILESCC(=O)OCC1=C(N2[C@H](SC1)[C@H](N)C2=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=95 %
Infrared spectrumConforms
Specific optical rotation+88° to +100° (20°C, 589 nm) (c=0.5, KH2PO4/NaOH)
Appearance (Color)Off-white to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only