Thermo Scientific Chemicals

3,3'-Dimethoxybenzidine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 184470050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3,3'-Dimethoxybenzidine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 184470050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 184470050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
20325-40-0
IUPAC Name
dihydrogen 3,3'-dimethoxy-[1,1'-biphenyl]-4,4'-diamine dichloride
Molecular Formula
C14H18Cl2N2O2
InChI Key
UXTIAFYTYOEQHV-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N)C1=CC=C(N)C(OC)=C1
Appearance (Color)
White to beige to grey to purple-grey
Titration with HClO4
>=97.5 % (on dry substance)
Appearance (Form)
Powder
Loss on drying
=<5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text