N-(2-Chloroethyl)acetamide, 98%, Thermo Scientific Chemicals
N-(2-Chloroethyl)acetamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(2-Chloroethyl)acetamide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 184520100
Price (USD)/ Each
43.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
50.80 
Save 7.15 (14%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
43.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
50.80 
Save 7.15 (14%)
Add to cart
N-(2-Chloroethyl)acetamide, 98%, Thermo Scientific Chemicals
Catalog number184520100
Price (USD)/ Each
43.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
50.80 
Save 7.15 (14%)
-
Add to cart
Chemical Identifiers
CAS7355-58-0
IUPAC NameN-(2-chloroethyl)acetamide
Molecular FormulaC4H8ClNO
InChI KeyHSKNJSHFPPHTAQ-UHFFFAOYSA-N
SMILESCC(=O)NCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Viscous liquid
Infrared spectrumConforms
Refractive index1.4770 to 1.4800 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only