Thermo Scientific Chemicals

3-Nitrobenzylamine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 184730050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Nitrobenzylamine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 184730050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 184730050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
26177-43-5
IUPAC Name
hydrogen 1-(3-nitrophenyl)methanamine chloride
Molecular Formula
C7H9ClN2O2
InChI Key
DLZXLCHQWOZGSE-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCC1=CC=CC(=C1)[N+]([O-])=O
Appearance (Color)
Off-white to tan
Infrared spectrum
Conforms
Appearance (Form)
Solid
Melting point
226°C to 231°C
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text