Thermo Scientific Chemicals

Hydroxypropyl cellulose, Average M.W. 100.000

Catalog number: 184880051
5 kg, Plastic drum, Each
Thermo Scientific Chemicals

Hydroxypropyl cellulose, Average M.W. 100.000

Catalog number: 184880051
5 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 184880051
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-64-2
Sulfated ash
=<0.5 %
Viscosity
75 to 150 mPa.s (5 % in water) (Brookfield settings: 30 rpm, spindle no.1)
Loss on drying
=<5 % (105°C)
Infrared spectrum
Conforms
Heavy metals (as Pb)
=<20 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text