Hydroxypropyl cellulose, Average M.W. 100.000
Hydroxypropyl cellulose, Average M.W. 100.000
Hydroxypropyl cellulose, Average M.W. 100.000
Hydroxypropyl cellulose, Average M.W. 100.000
Thermo Scientific Chemicals

Hydroxypropyl cellulose, Average M.W. 100.000

CAS: 9004-64-2 | (C24H44O16)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 184882500
Price (USD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9004-64-2
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.5 %
Viscosity75 to 150 mPa.s (5 % in water) (Brookfield settings: 30 rpm, spindle no.1)
Loss on drying=<5 % (105°C)
Infrared spectrumConforms
Heavy metals (as Pb)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link