Thermo Scientific Chemicals

Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aq.sol.; ca. MW 800,000, Thermo Scientific Chemicals

Catalog number: 184960250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aq.sol.; ca. MW 800,000, Thermo Scientific Chemicals

Catalog number: 184960250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 184960250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
27119-07-9
Molecular Formula
(C7H13NO4S)n
SMILES
CC(C)(CS(O)(=O)=O)NC(=O)C(-*)C-*
MDL Number
MFCD00084369
Assay
8.0 to 12.0 % Solids
Appearance (Form)
Clear viscous liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text