Thermo Scientific Chemicals

Poly(acrylic acid), 63 wt.% solution in water; approx. M.W. 2,000, Thermo Scientific Chemicals

Catalog number: 184990050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Poly(acrylic acid), 63 wt.% solution in water; approx. M.W. 2,000, Thermo Scientific Chemicals

Catalog number: 184990050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 184990050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9003-01-4
IUPAC Name
Poly(acrylic acid)
Molecular Formula
(C3H4O2)n
SMILES
OC(=O)C(-*)C-*
MDL Number
MFCD00084394
Infrared spectrum
Conforms
Assay
60.0 to 65.0 % Solids
Appearance (Form)
Clear to hazy liquid
Molecular weight
2000 (approx)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text