Poly(acrylic acid), 63 wt.% solution in water; approx. M.W. 2,000, Thermo Scientific Chemicals
Poly(acrylic acid), 63 wt.% solution in water; approx. M.W. 2,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(acrylic acid), 63 wt.% solution in water; approx. M.W. 2,000, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
250 g
Packaging:
Glass bottle
Catalog number 184995000
Price (USD)/ Each
167.65
Online exclusive
186.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
167.65
Online exclusive
186.00 
Save 18.35 (10%)
Add to cart
Poly(acrylic acid), 63 wt.% solution in water; approx. M.W. 2,000, Thermo Scientific Chemicals
Catalog number184995000
Price (USD)/ Each
167.65
Online exclusive
186.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9003-01-4
IUPAC NamePoly(acrylic acid)
Molecular Formula(C3H4O2)n
SMILESOC(=O)C(-*)C-*
MDL NumberMFCD00084394
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Assay60.0 to 65.0 % Solids
Appearance (Form)Clear to hazy liquid
Molecular weight2000 (approx)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only