Ethyl nitroacetate, 98%, Thermo Scientific Chemicals
Ethyl nitroacetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl nitroacetate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 185170050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
IUPAC Nameethyl 2-nitroacetate
Molecular FormulaC4H7NO4
InChI KeyFTKASJMIPSSXBP-UHFFFAOYSA-N
SMILESCCOC(=O)C[N+]([O-])=O
Molecular Weight (g/mol)133.10
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.4235 to 1.4255 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only