Ethyl nitroacetate, 98%, Thermo Scientific Chemicals
Ethyl nitroacetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl nitroacetate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 185170250
Price (USD)/ Each
171.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
202.00 
Save 30.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
171.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
202.00 
Save 30.35 (15%)
Add to cart
Ethyl nitroacetate, 98%, Thermo Scientific Chemicals
Catalog number185170250
Price (USD)/ Each
171.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
202.00 
Save 30.35 (15%)
-
Add to cart
Chemical Identifiers
CAS626-35-7
IUPAC Nameethyl 2-nitroacetate
Molecular FormulaC4H7NO4
InChI KeyFTKASJMIPSSXBP-UHFFFAOYSA-N
SMILESCCOC(=O)C[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.4235 to 1.4255 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only